3-(Methylthio)phenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid

224400-1G 224400-250MG 224400-5G
BT224400-1GEA 133 USD
BT224400-1G BT224400-250MG BT224400-5G
3-(Methylthio)phenylboronic acid ≥97%
3-(Methylthio)phenylboronic acid

CAS: 128312-11-8; MDL No: MFCD01319105 Solid; Molecular Formula: C7H9BO2S; MW: 168.02 Melting Point: 160-162°; Boiling Point: 210-215° Density (g/mL): 1.087


  • Solid

Formula: C₇H₉BO₂S
MW: 168.02 g/mol
Boiling Pt: 210…215 °C
Melting Pt: 160…162 °C
Density: 1.087 g/mL
MDL Number: MFCD01319105
CAS Number: 128312-11-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR