5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥99%

Supplier: BeanTown Chemical

Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid

224385-1G 224385-25G 224385-5G
BT224385-1GEA 34.2 USD
BT224385-1G BT224385-25G BT224385-5G
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥99%
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

CAS: 69-78-3; EC No: 200-714-4; MDL No: MFCD00007140; RTECS: DG9650000 Powder; Molecular Formula: C14H8N2O8S2; MW: 396.35 Melting Point: 240-245° (decomposes)


  • RTECS: DG9650000
  • Powder

Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
Melting Pt: 240…245 °C (dec.)
MDL Number: MFCD00007140
CAS Number: 69-78-3
EINECS: 200-714-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR