Formamidine disulfide dihydrochloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Dithiourea Dihydrochloride, Dithiodiformamide Dihydrochloride

224365-100G 224365-25G 224365-5G
BT224365-100GEA 694 USD
BT224365-100G BT224365-25G BT224365-5G
Formamidine disulfide dihydrochloride ≥97%
Formamidine disulfide dihydrochloride

CAS: 14807-75-1; EC No: 238-876-3; MDL No: MFCD00012586; RTECS: LQ4570000 Powder; Molecular Formula: CH7Cl2N2S2; MW: 223.15 Melting Point: 175-180° (decomposes) Hygroscopic


  • RTECS: LQ4570000
  • Hygroscopic
  • Powder

Formula: C₂H₆N₄S₂
MW: 223.13 g/mol
Melting Pt: 175…180 °C (dec.)
MDL Number: MFCD00012586
CAS Number: 14807-75-1
EINECS: 238-876-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR