N-(4-Oxocyclohexyl)acetamide ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Acetamidocyclohexanone

224360-100G 224360-25G 224360-5G
BT224360-100GEA 280 USD
BT224360-100G BT224360-25G BT224360-5G
N-(4-Oxocyclohexyl)acetamide ≥97%
N-(4-Oxocyclohexyl)acetamide

CAS: 27514-08-5; MDL No: MFCD03703462 Solid; Molecular Formula: C8H13NO2 ; MW: 155.19 Melting Point: 136-140°


  • Solid

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
Melting Pt: 136…140 °C
MDL Number: MFCD03703462
CAS Number: 27514-08-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR