Cyclohexen-1-ylbenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Phenylcyclohexene, 1-Phenyl-1-cyclohexene, 2,3,4,5-Tetrahydro-1,1'-biphenyl

224355-100G 224355-25G 224355-5G
BT224355-100GEA 498 USD
BT224355-100G BT224355-25G BT224355-5G
Cyclohexen-1-ylbenzene ≥97%
Cyclohexen-1-ylbenzene

CAS: 771-98-2; EC No: 212-242-6; MDL No: MFCD00001542; RTECS: GW6600000 Liquid; Molecular Formula: C12H14; MW: 158.24 Melting Point: -11°; Boiling Point: 251-253°; Flash point: 103°C (217°F) Density (g/mL): 0.994; Refractive Index: 1.570


  • RTECS: GW6600000
  • Refractive index: 1.570
  • Liquid

Formula: C₁₂H₁₄
MW: 158.24 g/mol
Boiling Pt: 251…253 °C
Melting Pt: –11 °C
Density: 0.994 g/mL
Flash Pt: 103 °C (217 °F)
MDL Number: MFCD00001542
CAS Number: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce31989847b7f5823ac4c86db68428fc148afb1d&vid=eeeb1ffa4c17dd4e58f5fc1f819153e435054e02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR