4,4'-Isopropylidenediphenol ≥97%

Supplier: BeanTown Chemical

224320-1KG 224320-250G 224320-5KG
BT224320-1KGEA 59.56 USD
BT224320-1KG BT224320-250G BT224320-5KG
4,4'-Isopropylidenediphenol ≥97%
4,4'-Isopropylidenediphenol

CAS: 80-05-7; EC No: 201-245-8; MDL No: MFCD00002366; RTECS: SL6300000 Crystalline; Molecular Formula: C15H16O2; MW: 228.29 Melting Point: 158-159°; Boiling Point: 200°/4 mmHg


  • RTECS: SL6300000
  • Crystalline

Formula: C₁₅H₁₆O₂
Boiling Pt: 200 °C/4 mmHg
Melting Pt: 158…159 °C
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR