2-(Difluoromethoxy)benzaldehyde ≥97%
Supplier: BeanTown Chemical
224305-1G
224305-5G
BT224305-1GEA
46
USD
BT224305-1G
BT224305-5G
2-(Difluoromethoxy)benzaldehyde ≥97%
2-(Difluoromethoxy)benzaldehyde
CAS: 71653-64-0; MDL No: MFCD00042251
Liquid; Molecular Formula: C8H6F2O2 ; MW: 172.13
Boiling Point: 216°; Flash point: 107°C (225°F)
Density (g/mL): 1.30; Refractive Index: 1.497
Air Sensitive
- Refractive index: 1.497
- Air Sensitive
- Liquid
Formula:
C₈H₆F₂O₂ Boiling Pt: 216 °C Density: 1.30 g/mL Flash Pt: 107 °C (225 °F) |
MDL Number:
MFCD00042251 CAS Number: 71653-64-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...