2,2-Dimethyldihydro-2H-pyran-4(3H)-one ≥95%
Supplier: BeanTown Chemical
Synonyms:
Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 2,2-Dimethyltetrahydropyran-4-one
224300-1G
224300-250MG
BT224300-1GEA
312
USD
BT224300-1G
BT224300-250MG
2,2-Dimethyldihydro-2H-pyran-4(3H)-one ≥95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one
CAS: 1194-16-7; MDL No: MFCD01549337
Liquid; Molecular Formula: C7H12O2; MW: 128.17
Boiling Point: 60-64°/8 mmHg
- Liquid
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 60…64 °C/8 mmHg |
MDL Number:
MFCD01549337 CAS Number: 1194-16-7 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...