3,5-Dibromo-2-fluoro-4-methylpyridine

Supplier: BeanTown Chemical

Synonyms: 2-Fluoro-3,5-dibromo-4-methylpyridine

224290-1G 224290-5G
BT224290-1GEA 208 USD
BT224290-1G BT224290-5G
3,5-Dibromo-2-fluoro-4-methylpyridine
3,5-Dibromo-2-fluoro-4-methylpyridine

CAS: 1000340-01-1; MDL No: MFCD09864703 Solid; Molecular Formula: C6H4NBr2F; MW: 268.91


  • Solid

Formula: C₆H₄NBr₂F
MW: 268.9100 g/mol
MDL Number: MFCD09864703
CAS Number: 1000340-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR