4'-Bromoacetophenone ≥98%

Supplier: BeanTown Chemical

Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

224250-100G 224250-25G 224250-500G
BT224250-100GEA 73.4 USD
BT224250-100G BT224250-25G BT224250-500G
4'-Bromoacetophenone ≥98%
4'-Bromoacetophenone

CAS: 99-90-1; EC No: 202-799-3; MDL No: MFCD00000105 Solid; Molecular Formula: C8H7BrO; MW: 199.04 Melting Point: 49-51°; Boiling Point: 255°; Flash point: 110°C (230°F)


  • Solid

Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 255 °C
Melting Pt: 49…51 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR