Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm
Supplier: BeanTown Chemical
224240-25X25MM
224240-50X50MM
BT224240-25X25EA
359
USD
BT224240-25X25
BT224240-50X50
Platinum iridium alloy (80:20 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm
Platinum iridium alloy (80:20 w%)
MDL No: MFCD02091683
Solid; Formula: Pt:Ir; 80:20 wt%
- Solid
MDL Number:
MFCD02091683 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...