Zincon sodium salt
Supplier: BeanTown Chemical
224205-1G
224205-25G
224205-5G
BT224205-1GEA
31.63
USD
BT224205-1G
BT224205-25G
BT224205-5G
Zincon sodium salt
Zincon sodium salt
CAS: 62625-22-3; EC No: 263-651-1; MDL No: MFCD00064385
Solid; Molecular Formula: C20H15N4NaO6S; MW: 462.42
Melting Point: 203° (decomposes)
- Solid
Formula:
C₂₀H₁₅N₄NaO₆S Melting Pt: 203 °C (dec.) |
MDL Number:
MFCD00064385 CAS Number: 62625-22-3 EINECS: 263-651-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...