2-Bromo-4-pyridineboronic acid pinacol ester ≥97%

Supplier: BeanTown Chemical

224180-1G 224180-250MG
BT224180-1GEA 323 USD
BT224180-1G BT224180-250MG
2-Bromo-4-pyridineboronic acid pinacol ester ≥97%
2-Bromo-4-pyridineboronic acid pinacol ester

CAS: 458532-82-6; MDL No: MFCD06798252 Crystalline/Powder; Molecular Formula: C11H15BBrNO2; MW: 283.96


  • Crystalline/Powder

Formula: C₁₁H₁₅BBrNO₂
MDL Number: MFCD06798252
CAS Number: 458532-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR