3-Ethynylaniline ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Amino-3-ethynylbenzene, (3-Aminophenyl)acetylene, (3-Aminophenyl)ethyne, 1-Ethynyl-3-aminobenzene

224170-25G 224170-5G
BT224170-25GEA 225 USD
BT224170-25G BT224170-5G
3-Ethynylaniline ≥98%
3-Ethynylaniline

CAS: 54060-30-9; EC No: 258-944-6; MDL No: MFCD00014779 Liquid; Molecular Formula: C8H7N; MW: 117.15 Density (g/mL): 1.04


  • Liquid

Formula: C₈H₇N
Density: 1.04 g/mL
MDL Number: MFCD00014779
CAS Number: 54060-30-9
EINECS: 258-944-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR