Rhodium(III) oxide, anhydrous ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

224160-2G 224160-500MG
BT224160-2GEA 2103 USD
BT224160-2G BT224160-500MG
Rhodium(III) oxide, anhydrous ≥99.9% (trace metals basis)
Rhodium (III) oxide

CAS: 12036-35-0; EC No: 234-846-9; MDL No: MFCD00011205 Powder; Linear Formula: Rh2O3; MW: 253.81 Melting Point: 1100° (decomposes) Density (g/mL): 8.2 Hygroscopic


  • Hygroscopic
  • Powder

Formula: O₃Rh₂
Melting Pt: 1100 °C (dec.)
Density: 8.2 g/mL
MDL Number: MFCD00011205
CAS Number: 12036-35-0
EINECS: 234-846-9

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR