TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥99%
Supplier: BeanTown Chemical
Synonyms:
TBTU
224140-1G
224140-25G
224140-5G
BT224140-1GEA
21
USD
BT224140-1G
BT224140-25G
BT224140-5G
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) ≥99%
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
CAS: 125700-67-6; EC No: 423-040-4; MDL No: MFCD00077413
Crystalline; Linear Formula: C11H16N5O·BF4; MW: 321.08
Melting Point: 205° (decomposes)
Light Sensitive, Moisture Sensitive
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₁₁H₁₆BF₄N₅O MW: 321.1 g/mol Melting Pt: 205 °C (dec.) |
MDL Number:
MFCD00077413 CAS Number: 125700-67-6 EINECS: 423-040-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...