4'-Methoxyacetophenone ≥99%
Supplier: BeanTown Chemical
CAS: 100-06-1; EC No: 202-815-9; MDL No: MFCD00008745; RTECS: AM9240000
Crystalline; Molecular Formula: C9H10O2; MW: 150.17
Melting Point: 36-38°; Boiling Point: 152-154°/26 mmHg; Flash point: 138°C (280°F)
Light Sensitive
- RTECS: AM9240000
- Light Sensitive
- Crystalline
Formula:
C₉H₁₀O₂ Boiling Pt: 152…154 °C/26 mmHg Melting Pt: 36…38 °C Flash Pt: 138 °C (280 °F) |
MDL Number:
MFCD00008745 CAS Number: 100-06-1 EINECS: 202-815-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...