2-Amino-5-bromo-4-methylpyridine ≥97%
Supplier: BeanTown Chemical
224085-1G
224085-25G
224085-5G
BT224085-1GEA
34
USD
BT224085-1G
BT224085-25G
BT224085-5G
2-Amino-5-bromo-4-methylpyridine ≥97%
2-Amino-5-bromo-4-methylpyridine
CAS: 98198-48-2; MDL No: MFCD03427660
Powder; Molecular Formula: C6H7BrN2; MW: 187.04
Melting Point: 148-151°
- Powder
Formula:
C₆H₇BrN₂ Melting Pt: 148…151 °C |
MDL Number:
MFCD03427660 CAS Number: 98198-48-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...