Azure II

Supplier: BeanTown Chemical
Warning

Synonyms: C.I. 52010/52015, 3,7 Bis(dimethylamino)phenothiazin-5-ium chloride and 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium chloride, Azur B/Methylene Blue 1:1, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride mixt. with 3-(dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride(1:1), Methylene Azure II

224070-10G 224070-50G
BT224070-10GEA 32 USD
BT224070-10G BT224070-50G
Azure II
Azure II

CAS: 37247-10-2; MDL No: MFCD00080716 Powder; Molecular Formula: C16H18N3S·C15H16N3S·2Cl; MW: 625.68


  • Powder

Formula: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
MW: 625.68 g/mol
MDL Number: MFCD00080716
CAS Number: 37247-10-2
Merck Index: 14,6058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR