Lauric acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dodecanoic acid, Dodecylic acid
224050-10KG
224050-2.5KG
224050-500G
BT224050-10KGEA
232
USD
BT224050-10KG
BT224050-2.5KG
BT224050-500G
Lauric acid ≥98%
Lauric acid
CAS: 143-07-7; EC No: 205-582-1; MDL No: MFCD00002736; RTECS: OE9800000
Crystalline; Linear Formula: CH3(CH2)10CO2H; Molecular Formula: C12H24O2; MW: 200.32
Melting Point: 44-46°; Boiling Point: 225°/100 mmHg; Flash point:
- RTECS: OE9800000
- Crystalline
Formula:
C₁₂H₂₄O₂ MW: 200.32 g/mol Boiling Pt: 225 °C/100 mmHg Melting Pt: 44…46 °C Density: 0.883 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00002736 CAS Number: 143-07-7 EINECS: 205-582-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...