Disodium oxalate ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
224030-1KG
224030-250G
224030-50G
BT224030-1KGEA
175
USD
BT224030-1KG
BT224030-250G
BT224030-50G
Disodium oxalate ≥99.5% ACS
Disodium oxalate
CAS: 62-76-0; EC No: 200-550-3; MDL No: MFCD00012465; RTECS: KI1750000
Solid; Linear Formula: Na2C2O4; MW: 134.00
Melting Point: 250-270°
Density (g/mL): 2.34
Hygroscopic
- RTECS: KI1750000
- Hygroscopic
- Solid
Formula:
Na₂C₂O₄ Melting Pt: 250…270 °C Density: 2.34 g/mL |
MDL Number:
MFCD00012465 CAS Number: 62-76-0 EINECS: 200-550-3 |
Specification Test Results
Assay | 99.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...