1-Acetamido-7-hydroxynaphthalene

Supplier: BeanTown Chemical

Synonyms: N-(7-Hydroxynaphthalen-1-yl)acetamide

223935-1G 223935-5G
BT223935-1GEA 216 USD
BT223935-1G BT223935-5G
1-Acetamido-7-hydroxynaphthalene
1-Acetamido-7-hydroxynaphthalene

CAS: 6470-18-4; EC No: 229-293-5; MDL No: MFCD00035710 Solid; Molecular Formula: C12H11NO2; MW: 201.22


  • Solid

Formula: C₁₂H₁₁NO₂
MW: 201.225 g/mol
MDL Number: MFCD00035710
CAS Number: 6470-18-4
EINECS: 229-293-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR