6-Amino-5-bromoquinoxaline ≥98%
Supplier: BeanTown Chemical
223900-1G
223900-5G
BT223900-1GEA
155
USD
BT223900-1G
BT223900-5G
6-Amino-5-bromoquinoxaline ≥98%
6-Amino-5-bromoquinoxaline
CAS: 50358-63-9; MDL No: MFCD00799024
Solid; Molecular Formula: C8H6BrN3; MW: 224.06
Melting Point: 151-153°
- Solid
Formula:
C₈H₆BrN₃ MW: 224.0573 g/mol Melting Pt: 151…153 °C |
MDL Number:
MFCD00799024 CAS Number: 50358-63-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...