(S)-(+)-Methyl mandelate ≥99%
Supplier: BeanTown Chemical
Synonyms:
L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate
223890-1G
223890-5G
BT223890-1GEA
32
USD
BT223890-1G
BT223890-5G
(S)-(+)-Methyl mandelate ≥99%
(S)-(+)-Methyl mandelate
CAS: 21210-43-5; MDL No: MFCD00064246
Powder; Molecular Formula: C9H10O3 ; MW: 166.17
Melting Point: 56-58°
Optical Rotation: [α]20/D +144°, c = 1 in methanol
- Optical rotation: [α]20/D +144°, c = 1 in methanol
- Powder
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 58 |
MDL Number:
MFCD00064246 CAS Number: 21210-43-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...