5-Bromo-2-methoxypyridine ≥98%

Supplier: BeanTown Chemical

223845-25G 223845-5G
BT223845-25GEA 194 USD
BT223845-25G BT223845-5G
5-Bromo-2-methoxypyridine ≥98%
5-Bromo-2-methoxypyridine

CAS: 13472-85-0; MDL No: MFCD01318952 Liquid; Molecular Formula: C6H6BrNO ; MW: 188.02 Boiling Point: 80°/12 mmHg; Flash point: 96°C (205°F) Density (g/mL): 1.453; Refractive Index: 1.555


  • Refractive index: 1.555
  • Liquid

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 80 °C/12 mmHg
Density: 1.453 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR