9,10-Bis(phenylethynyl)anthracene
Supplier: BeanTown Chemical
223835-1G
223835-25G
223835-5G
BT223835-1GEA
35.17
USD
BT223835-1G
BT223835-25G
BT223835-5G
9,10-Bis(phenylethynyl)anthracene
9,10-Bis(phenylethynyl)anthracene
CAS: 10075-85-1; EC No: 233-210-8; MDL No: MFCD00012050
Crystalline; Molecular Formula: C30H18; MW: 378.46
Melting Point: 248-250°
Light Sensitive
- Light Sensitive
- Crystalline
Formula:
C₃₀H₁₈ MW: 378.47 g/mol Melting Pt: 248…250 °C |
MDL Number:
MFCD00012050 CAS Number: 10075-85-1 EINECS: 233-210-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...