Ammonium dihydrogen phosphate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

223830-2KG 223830-500G
BT223830-2KGEA 149 USD
BT223830-2KG BT223830-500G
Ammonium dihydrogen phosphate ≥98.0% ACS
Ammonium dihydrogen phosphate

CAS: 7722-76-1; EC No: 231-764-5; MDL No: MFCD00003396 Crystalline; Linear Formula: NH4H2PO4; Molecular Formula: H6NO4P; MW: 115.03 Melting Point: 190° (decomposes)


  • Crystalline

Formula: NH₄H₂PO₄
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR