Di-tert-butyl malonate ≥98%

Supplier: BeanTown Chemical

223805-10G 223805-250G 223805-50G
BT223805-10GEA 62 USD
BT223805-10G BT223805-250G BT223805-50G
Di-tert-butyl malonate ≥98%
Di-tert-butyl malonate

CAS: 541-16-2; EC No: 208-769-6; MDL No: MFCD00008810 Liquid; Linear Formula: (CH3)3COCOCH2COOC(CH3)3; Molecular Formula: C11H20O4; MW: 216.28 Melting Point: -7 - -6°; Boiling Point: 110-111°/22 mmHg; Flash point: 89°C (192°F) Density (g/mL): 0.966; Refractive Index: 1.418


  • Refractive index: 1.418
  • Liquid

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 110…111 °C/22 mmHg
Melting Pt: –7 – –6 °C
Density: 0.966 g/mL
Flash Pt: 89 °C (192 °F)
MDL Number: MFCD00008810
CAS Number: 541-16-2
EINECS: 208-769-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR