Hexaammineruthenium(II) chloride ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

223760-1G 223760-250MG
BT223760-1GEA 186.86 USD
BT223760-1G BT223760-250MG
Hexaammineruthenium(II) chloride ≥99.9% (trace metals basis)
Hexaammineruthenium (II) chloride

CAS: 15305-72-3; EC No: 239-342-2; MDL No: MFCD00011477 Solid; Linear Formula: Ru(NH3)6Cl2 ; Molecular Formula: H18Cl2N6Ru; MW: 274.16


  • Solid

Formula: H₁₈Cl₂N₆Ru
MW: 274.16 g/mol
MDL Number: MFCD00011477
CAS Number: 15305-72-3
EINECS: 239-342-2

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR