2,6-Dichloro-3-nitropyridine ≥97%

Supplier: BeanTown Chemical

223745-10G 223745-250G 223745-50G
BT223745-10GEA 60 USD
BT223745-10G BT223745-250G BT223745-50G
2,6-Dichloro-3-nitropyridine ≥97%
2,6-Dichloro-3-nitropyridine

CAS: 16013-85-7; EC No: 240-151-1; MDL No: MFCD00006234 Powder; Molecular Formula: C5H2Cl2N2O2; MW: 192.99 Melting Point: 55-60°


  • Powder

Formula: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Melting Pt: 55…60 °C
MDL Number: MFCD00006234
CAS Number: 16013-85-7
EINECS: 240-151-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR