Bismuth (III) phosphate ≥99%
Supplier: BeanTown Chemical
223735-250G
223735-50G
BT223735-250GEA
353
USD
BT223735-250G
BT223735-50G
Bismuth (III) phosphate ≥99%
Bismuth (III) phosphate
CAS: 10049-01-1; EC No: 233-161-2; MDL No: MFCD00049721
Powder; Linear Formula: BiPO4; MW: 303.95
Melting Point:
- Powder
Formula:
BiO₄P MW: 303.95 g/mol Melting Pt: >350 °C Density: 6.32 g/mL |
MDL Number:
MFCD00049721 CAS Number: 10049-01-1 EINECS: 233-161-2 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...