(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride
223705-1G
223705-5G
BT223705-1GEA
76
USD
BT223705-1G
BT223705-5G
(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98%
(S)-2,4-Diaminobutanoic acid dihydrochloride
CAS: 1883-09-6; EC No: 217-542-0; MDL No: MFCD00064561
Crystalline/Solid; Molecular Formula: C4H11N2O2·2HCl; MW: 191.06
Melting Point: 197-200° (decomposes)
Optical Rotation: [α]20/D -14.5±1.5°, c = 3.67% in water
- Optical rotation: [α]20/D -14.5±1.5°, c = 3.67% in water
- Crystalline/Solid
Formula:
C₄H₁₂Cl₂N₂O₂ MW: 191.06 g/mol Melting Pt: 197…200 °C (dec.) |
MDL Number:
MFCD00064561 CAS Number: 1883-09-6 EINECS: 217-542-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...