1-Butyl-3-methylimidazoliumhexafluorophosphate ≥98%

Supplier: BeanTown Chemical

223685-100G 223685-25G 223685-5G
BT223685-100GEA 278 USD
BT223685-100G BT223685-25G BT223685-5G
1-Butyl-3-methylimidazoliumhexafluorophosphate ≥98%
1-Butyl-3-methylimidazoliumhexafluorophosphate

CAS: 174501-64-5; MDL No: MFCD03093295 Liquid; Molecular Formula: C8H15F6N2P; MW: 284.18 Density (g/mL): 1.38; Refractive Index: 1.410


  • Refractive index: 1.410
  • Liquid

Formula: C₈H₁₅F₆N₂P
Density: 1.38 g/mL
MDL Number: MFCD03093295
CAS Number: 174501-64-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR