Thioacetamide ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Ethanethioamide

223675-10G 223675-250G 223675-50G
BT223675-10GEA 25.15 USD
BT223675-10G BT223675-250G BT223675-50G
Thioacetamide ≥99.0% ACS
Thioacetamide

CAS: 62-55-5; EC No: 200-541-4; MDL No: MFCD00008070; RTECS: AC8925000 Crystalline; Linear Formula: CH3CSNH2; Molecular Formula: C2H5NS; MW: 75.13 Melting Point: 108-112°


  • RTECS: AC8925000
  • Crystalline

Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 108…112 °C
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR