4-Cyano-3-fluorophenyl-4-n-butylbenzoate ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Butylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Cyano-3-fluorophenyl 4-Butylbenzoate

223665-1G 223665-5G
BT223665-1GEA 103 USD
BT223665-1G BT223665-5G
4-Cyano-3-fluorophenyl-4-n-butylbenzoate ≥97%
4-Cyano-3-fluorophenyl-4-n-butylbenzoate

CAS: 86776-52-5; MDL No: MFCD11053362 Liquid; Molecular Formula: C18H16FNO2; MW: 297.33 Melting Point: 6.4-8.4°


  • Liquid

Formula: C₁₈H₁₆FNO₂
MW: 297.329 g/mol
Melting Pt: 6.4…8.4 °C
MDL Number: MFCD11053362
CAS Number: 86776-52-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR