4-Cyano-3-fluorophenyl-trans-4-(4-ethylcyclohexyl)benzoate ≥97%
Supplier: BeanTown Chemical
223655-1G
223655-5G
BT223655-1GEA
103
USD
BT223655-1G
BT223655-5G
4-Cyano-3-fluorophenyl-trans-4-(4-ethylcyclohexyl)benzoate ≥97%
4-Cyano-3-fluorophenyl-trans-4-(4-ethylcyclohexyl)benzoate
CAS: 92118-81-5; MDL No: MFCD11053364
Solid; Molecular Formula: C22H22FNO2; MW: 351.42
Melting Point: 89-90°
- Solid
Formula:
C₂₂H₂₂FNO₂ |
MDL Number:
MFCD11053364 CAS Number: 92118-81-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...