4-Methoxybenzyl mercaptan ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Methoxy-α-toluenethiol, p-Methoxytoluene-α-thiol
223650-25G
223650-5G
BT223650-25GEA
155
USD
BT223650-25G
BT223650-5G
4-Methoxybenzyl mercaptan ≥98%
4-Methoxybenzyl mercaptan
CAS: 6258-60-2; EC No: 228-393-6; MDL No: MFCD00004871
Liquid; Molecular Formula: C8H10OS; MW: 154.23
Boiling Point: 90-95°/0.5 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.107; Refractive Index: 1.573
Air Sensitive
- Refractive index: 1.573
- Air Sensitive
- Liquid
Formula:
C₈H₁₀OS MW: 154.23 g/mol Boiling Pt: 90…95 °C/0.5 mmHg Density: 1.107 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00004871 CAS Number: 6258-60-2 EINECS: 228-393-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...