2-Hydroxyphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxybenzeneboronic acid, Dihydroxy(2-hydroxyphenyl)borane, (2-Hydroxyphenyl)metaboric acid

223640-1G 223640-5G
BT223640-1GEA 74 USD
BT223640-1G BT223640-5G
2-Hydroxyphenylboronic acid ≥97%
2-Hydroxyphenylboronic acid

CAS: 89466-08-0; MDL No: MFCD01074581 Solid; Molecular Formula: C6H7BO3; MW: 137.93


  • Solid

Formula: C₆H₇BO₃
MW: 137.93 g/mol
MDL Number: MFCD01074581
CAS Number: 89466-08-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR