Indium (III) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Indium trioxide, Diindium trioxide
223625-10G
223625-250G
223625-50G
BT223625-10GEA
73.93
USD
BT223625-10G
BT223625-250G
BT223625-50G
Indium (III) oxide ≥99.99%
Indium (III) oxide
CAS: 1312-43-2; EC No: 215-193-9; MDL No: MFCD00011060
-325 Mesh Powder; Molecular Formula: In2O3; MW: 277.64
Density (g/mL): 7.18
- -325 Mesh Powder
Formula:
In₂O₃ MW: 277.64 g/mol Melting Pt: 715.05 Density: 7.18 g/mL |
MDL Number:
MFCD00011060 CAS Number: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...