3,4-Dichloro-2-nitro-6-(trifluoromethyl)toluene ≥98%

Supplier: BeanTown Chemical

223620-1G 223620-5G
BT223620-1GEA 31 USD
BT223620-1G BT223620-5G
3,4-Dichloro-2-nitro-6-(trifluoromethyl)toluene ≥98%
3,4-Dichloro-2-nitro-6-(trifluoromethyl)toluene

CAS: 115571-66-9; MDL No: MFCD04972788 Solid; Molecular Formula: C8H4NF3Cl2O2; MW: 274.02


  • Solid

Formula: C₈H₄Cl₂F₃NO₂
MDL Number: MFCD04972788
CAS Number: 115571-66-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR