Cesium acetate ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Acetic acid cesium salt

223615-100G 223615-25G
BT223615-100GEA 369 USD
BT223615-100G BT223615-25G
Cesium acetate ≥99.995%
Cesium acetate

CAS: 3396-11-0; EC No: 222-248-0; MDL No: MFCD00013056 Powder/Lump; Linear Formula: CsOOCCH3; Molecular Formula: C2H3CsO2; MW: 191.95 Melting Point: 194° Hygroscopic


  • Hygroscopic
  • Powder/Lump

Formula: H₃CCOOCs
MW: 191.95 g/mol
Melting Pt: 194
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR