Ammonium cerium (IV) sulfate dihydrate ≥94.0% ACS

Supplier: BeanTown Chemical

223590-1KG 223590-250G 223590-5KG
BT223590-1KGEA 200 USD
BT223590-1KG BT223590-250G BT223590-5KG
Ammonium cerium (IV) sulfate dihydrate ≥94.0% ACS
Ammonium cerium(IV) sulphate dihydrate

CAS: 10378-47-9; EC No: 231-567-4; MDL No: MFCD00149417 Crystalline; Linear Formula: (NH4)4Ce(SO4)4·2H2O; MW: 632.56 Melting Point: 130°


  • Crystalline

Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
Melting Pt: 130 °C
MDL Number: MFCD00149417
CAS Number: 10378-47-9
EINECS: 231-567-4

Order Now

Specification Test Results

Assay 94.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR