Carnauba wax, yellow No. 1

Supplier: BeanTown Chemical

223580-1KG 223580-250G 223580-5KG
BT223580-1KGEA 67 USD
BT223580-1KG BT223580-250G BT223580-5KG
Carnauba wax, yellow No. 1
Carnauba wax

CAS: 8015-86-9; EC No: 232-399-4; MDL No: MFCD00130724 Flakes Refractive Index: 1.45


  • Refractive index: 1.45
  • Flakes

MDL Number: MFCD00130724
CAS Number: 8015-86-9
EINECS: 232-399-4
Merck Index: 14,1846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR