Ammonium iron (III) sulfate dodecahydrate 95.8-102.0% ACS
Supplier: BeanTown Chemical
223570-2.5KG
223570-500G
BT223570-2.5KGEA
228
USD
BT223570-2.5KG
BT223570-500G
Ammonium iron (III) sulfate dodecahydrate 95.8-102.0% ACS
Ammonium iron(III) sulphate dodecahydrate
CAS: 7783-83-7; EC No: 233-382-4; MDL No: MFCD00150004; RTECS: WS5900000
Broken Crystals ; Linear Formula: NH4Fe(SO4)2·12H2O; MW: 482.19
Melting Point: 39-41°
- RTECS: WS5900000
- Broken Crystals
Formula:
NH₄Fe(SO₄)₂·12H₂O Melting Pt: 39…41 °C |
MDL Number:
MFCD00150004 CAS Number: 7783-83-7 EINECS: 233-382-4 |
Specification Test Results
Assay | 95.8-102.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...