(+)-Dibenzoyl-D-tartaric acid, anhydrous ≥99%

Supplier: BeanTown Chemical

Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid

223550-10G 223550-250G 223550-50G
BT223550-10GEA 26.61 USD
BT223550-10G BT223550-250G BT223550-50G
(+)-Dibenzoyl-D-tartaric acid, anhydrous ≥99%
(+)-Dibenzoyl-D-tartaric acid

CAS: 17026-42-5; EC No: 241-097-1; MDL No: MFCD00063222 Crystalline/Powder; Molecular Formula: C18H14O8; MW: 358.31 Melting Point: 154-156° Optical Rotation: [α]20/D +117°, c = 5% in ethanol Hygroscopic


  • Optical rotation: [α]20/D +117°, c = 5% in ethanol
  • Hygroscopic
  • Crystalline/Powder

Formula: C₁₈H₁₄O₈
Melting Pt: 154…156 °C
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR