3,4,7,8-Tetramethyl-1,10-phenanthroline ≥98%
Supplier: BeanTown Chemical
223540-1G
223540-5G
BT223540-1GEA
76.28
USD
BT223540-1G
BT223540-5G
3,4,7,8-Tetramethyl-1,10-phenanthroline ≥98%
3,4,7,8-Tetramethyl-1,10-phenanthroline
CAS: 1660-93-1; EC No: 216-762-4; MDL No: MFCD00004974
Powder; Molecular Formula: C16H16N2; MW: 236.31
Melting Point: 277-280°
- Powder
Formula:
C₁₆H₁₆N₂ MW: 236.32 g/mol Boiling Pt: 386 Melting Pt: 277…280 °C |
MDL Number:
MFCD00004974 CAS Number: 1660-93-1 EINECS: 216-762-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...