Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water

Supplier: BeanTown Chemical

223470-1G 223470-5G
BT223470-1GEA 52.34 USD
BT223470-1G BT223470-5G
Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water
Palladium hydroxide

CAS: 12135-22-7; EC No: 235-219-2; MDL No: MFCD00064599 Solid; Linear Formula: Pd(OH)2/C


  • Solid

Formula: H₂O₂Pd
MW: 140.435 g/mol
MDL Number: MFCD00064599
CAS Number: 12135-22-7
EINECS: 235-219-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR