Sodium methacrylate ≥95%
Supplier: BeanTown Chemical
223465-250G
223465-50G
BT223465-250GEA
173
USD
BT223465-250G
BT223465-50G
Sodium methacrylate ≥95%
Sodium methacrylate
CAS: 5536-61-8; EC No: 226-896-5; MDL No: MFCD00045886
Solid; Linear Formula: H2C=C(CH3)COONa; Molecular Formula: C4H5NaO2; MW: 108.07
Melting Point: 300°
Heat Sensitive, Moisture Sensitive
- Heat Sensitive, Moisture Sensitive
- Solid
Formula:
C₄H₅NaO₂ MW: 108.072 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00045886 CAS Number: 5536-61-8 EINECS: 226-896-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...