(4-Hydroxy-3-nitrophenyl)acetic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

223280-1G 223280-25G 223280-5G
BT223280-1GEA 44 USD
BT223280-1G BT223280-25G BT223280-5G
(4-Hydroxy-3-nitrophenyl)acetic acid ≥99%
(4-Hydroxy-3-nitrophenyl)acetic acid

CAS: 10463-20-4; EC No: 233-948-0; MDL No: MFCD00007122 Powder; Molecular Formula: C8H7NO5; MW: 197.14 Melting Point: 146-148°


  • Powder

Formula: (C₆H₃(OH)(NO₂))CH₂COOH
MW: 197.15 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD00007122
CAS Number: 10463-20-4
EINECS: 233-948-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR