Guaiacol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol

223270-1KG 223270-250G 223270-5KG
BT223270-1KGEA 86 USD
BT223270-1KG BT223270-250G BT223270-5KG
Guaiacol ≥98%
Guaiacol

CAS: 90-05-1; EC No: 201-964-7; MDL No: MFCD00002185; RTECS: SL7525000 Crystalline; Molecular Formula: C7H8O2; MW: 124.14 Melting Point: 26-29°; Boiling Point: 205°; Flash point: 82°C (179°F) Density (g/mL): 1.129; Refractive Index: 1.543 Air Sensitive, Light Sensitive


  • RTECS: SL7525000
  • Refractive index: 1.543
  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 205 °C
Melting Pt: 26…29 °C
Density: 1.129 g/mL
Flash Pt: 82 °C (179 °F)
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR